Få mere at vide
Veliparib, MedChemExpress
Artikelnummer. 16466047
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
200 mg
Pakningsstørrelse:
200mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16466047 5 mg -
16450926 200 mg 200mg
16443328 10 mg -
16414576 100 mg -
16409866 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16466047 Leverandør Medchem Express Leverandørnr. HY10129
MedChemExpress Veliparib (ABT-888) is a potent PARP inhibitor, inhibiting PARP1 and PARP2 with Kis of 5.2 and 2.9 nM, respectively.
Veliparib (ABT-888) is a potent PARP inhibitor, inhibiting PARP1 and PARP2 with Kis of 5.2 and 2.9 nM, respectively.
Tekniske data
Kemisk navn eller materiale Veliparib
Fysisk form Solid
CAS 912444-00-9
Farve Off-White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C13H16N4O
Til brug med (applikation) Cancer-programmed cell death
Synonym ABT-888
Opløselighedsinformation DMSO : ≥ 29 mg/mL (118.71 mM)
SMIL O=C(C1=C2NC([C@@]3(NCCC3)C)=NC2=CC=C1)N
Molekylvægt (g/mol) 244.29
Formel vægt 244.29
Procent renhed 98.0%