missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFR1/R2/R3 Antibody (FLT1/1663), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14285JF646
Additional Details : Weight : 0.00970kg
Description
VEGFR1/R2/R3 Monoclonal antibody specifically detects VEGFR1/R2/R3 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
VEGFR1/R2/R3 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Recombinant human VEGFR1/R2/R3 protein (Uniprot: P17948) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunofluorescence | |
FLT1/1663 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A or G purified | |
RUO | |
2321 | |
Store at 4°C in the dark. | |
Purified |