Få mere at vide
Vedaprofen-d3, TRC
Artikelnummer. 30450813
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30450813 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30450813 Leverandør Toronto Research Chemicals Leverandørnr. TRCV10530210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Vedaprofen-d3
CAS 1185054-34-5
Molekylær formel C19H19D3O2
Mængde 10 mg
Synonym 4-Cyclohexyl-alpha-(methyl-d3)-1-naphthaleneacetic Acid, 2-(4-Cyclohexil-1-naphthyl)-propionic Acid-d3, CERM 10202-d3, PM-150-d3, Quadrisol-d3
InChI formel InChI=1S/C19H22O2/c1-13(19(20)21)15-11-12-16(14-7-3-2-4-8-14)18-10-6-5-9-17(15)18/h5-6,9-14H,2-4,7-8H2,1H3,(H,20,21)/i1D3
SMIL [2H]C([2H])([2H])C(C(=O)O)c1ccc(C2CCCCC2)c3ccccc13
IUPAC navn 2-(4-cyclohexylnaphthalen-1-yl)-3,3,3-trideuteriopropanoic acid
Molekylvægt (g/mol) 285.4
Formel vægt 285.1808