Få mere at vide
Vecuronium Bromide, Mikromol™
Beskrivelse
Vecuronium Bromide
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Vecuronium bromide |
| CAS | 50700-72-6 |
| Analyt- eller komponentnavne | Vecuronium bromide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C34 H57 N2 O4 . Br |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Vecuronium bromide, Androstane, piperidinium deriv., Musculax, NC 45, Norcuron, Org-NC 45, Vecuronium, 1-[3alpha,17beta-Bis(acetyloxy)-2beta-(piperidin-1-yl)-5alpha-androstan-16beta-yl]-1-methylpiperidinium bromide, Piperidinium, 1-[(2beta,3alpha,5alpha,16beta,17beta)-3,17-bis(acetyloxy)-2-(1-piperidinyl)androstan-16-yl]-1-methyl-, bromide (1:1), Piperidinium, 1-[(2beta,3alpha,5alpha,16beta,17beta)-3,17-bis(acetyloxy)-2-(1-piperidinyl)androstan-16-yl]-1-methyl-, bromide (9CI), Pancuronium Bromide Impurity D as Bromide, Pancuronium Impurity D as Bromide |
| InChI formel | InChI=1S/C34H57N2O4.BrH/c1-23(37)39-31-20-25-12-13-26-27(34(25,4)22-29(31)35-16-8-6-9-17-35)14-15-33(3)28(26)21-30(32(33)40-24(2)38)36(5)18-10-7-11-19-36;/h25-32H,6-22H2,1-5H3;1H/q+1;/p-1/t25-,26+,27-,28-,29-,30-,31-,32-,33-,34-;/m0./s1 |
| SMIL | [Br-].CC(=O)O[C@H]1C[C@@H]2CC[C@@H]3[C@H](CC[C@@]4(C)[C@H]3C[C@@H]([C@@H]4OC(=O)C)[N+]5(C)CCCCC5)[C@@]2(C)C[C@@H]1N6CCCCC6 |
| IUPAC navn | [(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-17-acetyloxy-10,13-dimethyl-16-(1-methylpiperidin-1-ium-1-yl)-2-piperidin-1-yl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate;bromide |
| Vis mere |