Få mere at vide
Sonics Materials VCX-1500HV Ultrasonic Processor
Artikelnummer. 11783649
Skift visning
Watt:
1500 W
Pakningsstørrelse:
Hver
Artikelnummer. Watt Pakningsstørrelse
11783649 1500 W Hver
1 options
Artikelnummer. 11783649 Leverandør Sonics Materials Leverandørnr. VCX1500HV220
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Designed primarily for dispersing and homogenizing, this 1500 Watt high volume ultrasonic liquid processing system is ideal for large volume applications.
  • Ultrasonic processors and equipment for large volume applications
  • The flow cell throughput rate is typically 100 liters/hour – variables being viscosity and desired degree of processing
  • The flow cell is recommended for the treatment of low viscosity samples which do not require extended exposure to ultrasonics
  • Designed primarily for dispersing and homogenizing
  • The flow cell is easily disassembled for inspection and cleaning, and is water jacketed to enable the sample to be cooled/heated while it is being processed
Tekniske data
Watt 1500 W