Få mere at vide
Sonics Materials VC 750 Ultrasonic Processor
Artikelnummer. 11743619
Skift visning
Watt:
750 W
Pakningsstørrelse:
Hver
Artikelnummer. Watt Pakningsstørrelse
11743619 750 W Hver
1 options
Artikelnummer. 11743619 Leverandør Sonics Materials Leverandørnr. VC750220
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Robust and reliable 750 Watt ultrasonic processors for small and medium volume applications that are ideal for safely processing a wide range of organic and inorganic materials from 250 microliters to 1 liter.
  • Ultrasonic processors for small and medium volume applications
  • 750 watt ultrasonic processors to safely process a wide range of organic and inorganic materials, from 250 microliters to 1 liter
  • Typical applications include nanotechnology (producing nanoparticle materials and Graphene dispersions), cell lysing, sample preparation, homogenization, ChIP Assay, emulsification, disaggregation and deagglomeration, plus uses in the field of sonochemical liquid processing
  • Energy monitor, digital wattmeter, microprocessor-based and programmable
  • 10 hour process timer
Tekniske data
Produkttype Ultrasonic Processor
Watt 750 W