Få mere at vide
Tocris Bioscience™ Vatalanib succinate
Potent VEGFR inhibitor; also aromatase inhibitor
Brand: Tocris Bioscience™ 5680/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Vatalanib succinate is a potent VEGFR inhibitor (IC50 values are 37 and 77 nM for VEGFR-2 and -1, respectively). Inhibits proliferation, migration and survival of HUVECs in vitro and inhibits growth, vascularization and metastasis of tumors expressing VEGFR in mouse models. Also inhibits PDGFR-β, c-Kit and c-Fms. Potent aromatase inhibitor (IC50 = 50 nM). Orally available.Tekniske data
| N-(4-Chlorophenyl)-4-(4-pyridinylmethyl)-1-phthalazinamine succinate | |
| 212142-18-2 | |
| CGP 79787D, PTK787/ZK222584 | |
| 50 mg |