Få mere at vide
Vatalanib-d4 Dihydrochloride, TRC
Artikelnummer. 30453872
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30453872 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453872 Leverandør Toronto Research Chemicals Leverandørnr. TRCV09950210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Vatalanib-d4 Dihydrochloride
CAS 1246820-27-8
Molekylær formel C20H13D4Cl3N4
Mængde 10 mg
Synonym N-(4-Chlorophenyl-d4)-4-(4-pyridinylmethyl)-1-phthalazinamine Dihydrochloride, CGP-79787-d4
InChI formel InChI=1S/C20H15ClN4/c21-15-5-7-16(8-6-15)23-20-18-4-2-1-3-17(18)19(24-25-20)13-14-9-11-22-12-10-14/h1-12H,13H2,(H,23,25)/i5D,6D,7D,8D
SMIL ClC(C([2H])=C1[2H])=C([2H])C([2H])=C1NC2=NN=C(CC3=CC=NC=C3)C4=CC=CC=C42
Molekylvægt (g/mol) 423.76