missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Varicella Zoster Virus Glycoprotein E Antibody (15) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26891JF585
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Varicella Zoster Virus Glycoprotein E Monoclonal antibody specifically detects Varicella Zoster Virus Glycoprotein E in Virus samples. It is validated for ELISA,Western Blot
Tekniske data
Varicella Zoster Virus Glycoprotein E | |
Monoclonal | |
Janelia Fluor 585 | |
Envelope glycoprotein E, GE, ORF68 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Varicella Zoster Virus Glycoprotein E (Uniprot#: Q9J3M8; Met1-Tyr538). | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
ELISA, Western Blot | |
15 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Varicella Zoster Virus Glycoprotein E Antibody (15) [Janelia Fluor™ 585], Novus Biologicals™ > 0.1 mL; Janelia Fluor 585
Ser du en mulighed for forbedring?Del en indholdskorrektion