Learn More
WTW™ Variable Piston Pipettes
For use in photometric water analysis in the field and laboratory.
Brand: WTW™ 250546
3529.03 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manually adjustable and robust
- Provides high test throughput
- Fixed volume for quick handling
- Available in different volumes and steps
Specifications
Piston Lift Pipette | |
0.5 to 5 mL | |
Each |
Product Content Correction
Product Title
WTW™ Variable Piston Pipettes > Volume: 0.5 to 5mL; 10uL steps
Spot an opportunity for improvement?Share a Content Correction