Få mere at vide
Vanillylmandelic Acid-d3 Ethyl Ester, TRC
Artikelnummer. 30450716
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30450716 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30450716 Leverandør Toronto Research Chemicals Leverandørnr. TRCV097617100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Vanillylmandelic Acid-d3 Ethyl Ester
CAS 1329499-52-6
Molekylær formel C11H11D3O5
Mængde 100 mg
Synonym 4-Hydroxy-3-methoxy-mandelic Acid-d3 Ethyl Ester, α,4-Dihydroxy-3-methoxy-benzeneacetic Acid-d3 Ethyl Ester
InChI formel InChI=1S/C11H14O5/c1-3-16-11(14)10(13)7-4-5-8(12)9(6-7)15-2/h4-6,10,12-13H,3H2,1-2H3/i2D3
SMIL OC1=CC=C(C(C(OCC)=O)O)C=C1OC([2H])([2H])[2H]
Molekylvægt (g/mol) 229.24