Få mere at vide
Vanillylamine-d3 Hydrochloride, TRC
Artikelnummer. 30453168
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30453168 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453168 Leverandør Toronto Research Chemicals Leverandørnr. TRCV0976021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Vanillylamine-d3 Hydrochloride
CAS 1217899-51-8
Molekylær formel C8H9D3ClNO2
Mængde 1 mg
Synonym 4-(Aminomethyl)-2-methoxyphenol-d3 Hydrochloride, 4-Hydroxy-3-methoxybenzylamine-d3 Hydrochloride, 3-Methoxy-d3-4-hydroxybenzylamine Hydrochloride, alpha-Aminocreosol-d3 Hydrochloride
InChI formel InChI=1S/C8H11NO2.ClH/c1-11-8-4-6(5-9)2-3-7(8)10;/h2-4,10H,5,9H2,1H3;1H/i1D3;
SMIL Cl.[2H]C([2H])([2H])Oc1cc(CN)ccc1O
IUPAC navn 4-(aminomethyl)-2-(trideuteriomethoxy)phenol;hydrochloride
Molekylvægt (g/mol) 192.66
Formel vægt 192.0745