missing translation for 'onlineSavingsMsg'
Få mere at vide

Vanillyl Methyl Ketone-d5, TRC

Artikelnummer. 30449203
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30449203 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30449203 Supplier Toronto Research Chemicals Supplier No. TRCV09762710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Vanillyl Methyl Ketone-d5
CAS 1794811-86-1
Molekylær formel C10H7D5O3
Mængde 10 mg
Synonym 1-(4-Hydroxy-3-methoxyphenyl)-2-propanone-d5, (4-Hydroxy-3-methoxyphenyl)acetone-d5, 1-(4-Hydroxy-3-methoxyphenyl)-2-propanone-d5, 2-Propiovanillone-d5, Guaiacylacetone-d5, Methyl Vanillyl Ketone-d5, NSC 16690-d5
InChI formel InChI=1S/C10H12O3/c1-7(11)5-8-3-4-9(12)10(6-8)13-2/h3-4,6,12H,5H2,1-2H3/i1D3,5D2
SMIL [2H]C([2H])([2H])C(=O)C([2H])([2H])c1ccc(O)c(OC)c1
IUPAC navn 1,1,1,3,3-pentadeuterio-3-(4-hydroxy-3-methoxyphenyl)propan-2-one
Molekylvægt (g/mol) 185.23
Formel vægt 185.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.