Learn More
Vanadium for AAS 1g/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical B065500
Additional Details : Weight : 0.50000kg
Packaging | Bottle |
---|---|
Quantity | 500mL |
Chemical Identifiers
7697-37-2 | |
63.012 | |
944 | |
nitric acid |
HNO3 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
CHEBI:48107 | |
[N+](=O)(O)[O-] |
Specifications
Vanadium | |
HNO3 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
Bottle | |
Liquid | |
Atomic Absorption Spectroscopy Flame Single-Element Standard Solution |
7697-37-2 | |
1g/L ±0.002g/L | |
[N+](=O)(O)[O-] | |
63.012 | |
CHEBI:48107 | |
2% HNO3 | |
500mL |