Få mere at vide
Valsartan-d3, TRC
Artikelnummer. 30455463
Change view
Click to view available options
Mængde:
1 mg
Product Code. Mængde
30455463 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30455463 Supplier Toronto Research Chemicals Supplier No. TRCV0957521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemisk navn eller materiale Valsartan-d3
CAS 1331908-02-1
Molekylær formel C24 H26 D3 N5 O3
Mængde 1 mg
Synonym Valsartan D3 (Pentanoyl 5,5,5 D3), N-(1-Oxopentyl-5,5,5-d3)-N-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-L-valine, (2S)-3-Methyl-2-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl-(5,5,5-trideuteriopentanoyl)amino]butanoic acid, Valsartan-d3, Valsartan-D3 (pentanoyl-5,5,5-D3), (S)-(-)-Valsartan-d3 (ω-valeroyl-d3)
InChI formel InChI=1S/C24H29N5O3/c1-4-5-10-21(30)29(22(16(2)3)24(31)32)15-17-11-13-18(14-12-17)19-8-6-7-9-20(19)23-25-27-28-26-23/h6-9,11-14,16,22H,4-5,10,15H2,1-3H3,(H,31,32)(H,25,26,27,28)/t22-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])CCCC(=O)N(Cc1ccc(cc1)c2ccccc2c3nnn[nH]3)[C@@H](C(C)C)C(=O)O
IUPAC navn (2S)-3-methyl-2-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl-(5,5,5-trideuteriopentanoyl)amino]butanoic acid
Molekylvægt (g/mol) 438.542
Formel vægt 438.2459