Få mere at vide
Valproic Acid Methyl Ester, TRC
Artikelnummer. 30133172
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133172 10 g
30012541 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133172 Leverandør Toronto Research Chemicals Leverandørnr. TRCV09477510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22632-59-3
Molekylær formel C9 H18 O2
Molekylvægt (g/mol) 158.24
Synonym 2-Propylpentanoic acid methyl ester, Valeric acid, 2-propyl-, methyl ester (6CI,7CI,8CI), Methyl 2-propylpentanoate, Methyl dipropylacetate, Methyl valproate
IUPAC navn methyl 2-propylpentanoate
SMIL CCCC(CCC)C(=O)OC
Tekniske data
Kemisk navn eller materiale Methyl Valproate
Mængde 10 g
InChI formel InChI=1S/C9H18O2/c1-4-6-8(7-5-2)9(10)11-3/h8H,4-7H2,1-3H3
Formel vægt 158.1307