Få mere at vide
Valproic Acid-d4, TRC
Artikelnummer. 30451836
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30451836 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451836 Leverandør Toronto Research Chemicals Leverandørnr. TRCV0947531MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Valproic acid 4,4,4',4'-d4
CAS 87745-17-3
Molekylær formel C8 D4 H12 O2
Mængde 1 mg
Synonym Pentanoic-3,3-d2 acid','2-(propyl-1,1-d2)- (9CI), 3,3,3',3'-Tetradeuterovalproic acid, 2-(Propyl-1,1-d2)pentanoic-3,3-d2 acid, Valproic acid-d4, Valproic-3,3,3',3'-D4 acid
InChI formel InChI=1S/C8H16O2/c1-3-5-7(6-4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)/i3D2,4D2
SMIL [2H]C([2H])(C)CC(CC([2H])([2H])C)C(=O)O
IUPAC navn 4,4-dideuterio-2-(2,2-dideuteriopropyl)pentanoic acid
Molekylvægt (g/mol) 148.24
Formel vægt 148.1401