Få mere at vide
Valnoctamide-d5, TRC
Artikelnummer. 30456645
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30456645 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30456645 Leverandør Toronto Research Chemicals Leverandørnr. TRCV09450210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Valnoctamide-d5
CAS 1190015-82-7
Molekylær formel C8H12D5NO
Mængde 10 mg
Synonym 2-Ethyl-d5-3-methylpentan-amide, 2-Ethyl-d5-3-methylvaleramide, Valmethamide-d5, McN-X-181-d5, Axiquel-d5, Nirvanil-d5
InChI formel InChI=1S/C8H17NO/c1-4-6(3)7(5-2)8(9)10/h6-7H,4-5H2,1-3H3,(H2,9,10)/i2D3,5D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C(C(C)CC)C(=O)N
IUPAC navn 3-methyl-2-(1,1,2,2,2-pentadeuterioethyl)pentanamide
Molekylvægt (g/mol) 148.26
Formel vægt 148.1624