Få mere at vide
Valnemulin Trifluoroacetic Acid Salt-d6, TRC
Artikelnummer. 30449134
Skift visning
Mængde:
0.25 mg
Artikelnummer. Mængde
30449134 0.25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30449134 Leverandør Toronto Research Chemicals Leverandørnr. TRCV0944020.25MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Valnemulin-d6 trifluoroacetic acid salt
CAS 1217627-44-5
Renhedsgrad noter HPLC
Molekylær formel C33 H47 D6 F3 N2 O7 S
Mængde 0.25 mg
InChI formel InChI=1S/C31H52N2O5S/c1-10-29(8)15-22(38-23(35)16-39-28(6,7)17-33-27(37)24(32)18(2)3)30(9)19(4)11-13-31(20(5)26(29)36)14-12-21(34)25(30)31/h10,18-20,22,24-26,36H,1,11-17,32H2,2-9H3,(H,33,37)/t19?,20-,22+,24+,25-,26-,29+,30?,31-/m0/s1/i6D3,7D3
SMIL [2H]C([2H])([2H])C(CNC(=O)[C@H](N)C(C)C)(SCC(=O)O[C@@H]1C[C@@](C)(C=C)[C@@H](O)[C@H](C)[C@@]23CCC(C)C1(C)[C@@H]2C(=O)CC3)C([2H])([2H])[2H]
Molekylvægt (g/mol) 667.87
Formel vægt 570.3974
Procent renhed >95