Få mere at vide
Valethamate Bromide, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Valethamate Bromide |
| CAS | 90-22-2 |
| Analyt- eller komponentnavne | Valethamate Bromide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H32 N O2 . Br |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Ethanaminium, N,N-diethyl-N-methyl-2-[(3-methyl-1-oxo-2-phenylpentyl)oxy]-, bromide (1:1), Ammonium, diethyl(2-hydroxyethyl)methyl-, bromide, 3-methyl-2-phenylvalerate (8CI), Diethyl(2-hydroxyethyl)methylammonium bromide 3-methyl-2-phenylvalerate (6CI,7CI), Ethanaminium, N,N-diethyl-N-methyl-2-[(3-methyl-1-oxo-2-phenylpentyl)oxy]-, bromide (9CI), Valeric acid, 3-methyl-2-phenyl-, ester with diethyl(2-hydroxyethyl)methylammonium bromide (8CI), 2-Diethylaminoethyl 2-phenyl-3-methylvalerate methyl bromide, 2-Diethylaminoethyl 3-methyl-2-phenylvalerate methylbromide, 3-Methyl-2-phenylvaleric acid 2-diethylaminoethyl ester methyl bromide, 3-Methyl-2-phenylvaleric acid diethyl(2-hydroxyethyl)methylammonium bromide ester, Barespan, Diethyl(2-hydroxyethyl)methylammonium 3-methyl-2-phenylvalerate bromide, Elist, Epidosan, Epidosin, Letamate, Murel, Release V, Resitan, S 78, Valethamate bromide, Velamate, 2-Phenyl-3-methylvaleric acid beta-(diethylamino)ethyl ester bromomethylate |
| InChI formel | InChI=1S/C19H32NO2.BrH/c1-6-16(4)18(17-12-10-9-11-13-17)19(21)22-15-14-20(5,7-2)8-3;/h9-13,16,18H,6-8,14-15H2,1-5H3;1H/q+1;/p-1 |
| Løsningstype | Neat |
| SMIL | [Br-].CCC(C)C(C(=O)OCC[N+](C)(CC)CC)c1ccccc1 |
| Vis mere |