Få mere at vide
Valethamate bromide, 99%, Thermo Scientific™
376.41 DKK - 620.11 DKK
Kemiske identifikatorer
| CAS | 90-22-2 |
|---|---|
| Molekylær formel | C19H32BrNO2 |
| Molekylvægt (g/mol) | 386.37 |
| MDL nummer | MFCD00031617 |
| InChI nøgle | CEJGGHKJHDHLAZ-UHFFFAOYNA-M |
| Synonym | valethamate bromide, ediposin, epidosin, epidozin, resitan, murel, valethamate bromide jan, n,n-diethyl-n-methyl-2-3-methyl-2-phenylpentanoyl oxy ethanaminium bromide, diethyl methyl 2-3-methyl-2-phenylvaleryloxy ethyl ammonium bromide, 2-diethylaminoethyl 2-phenyl-3-methylvalerate methyl bromide |
| PubChem CID | 7010 |
| IUPAC navn | diethyl(methyl){2-[(3-methyl-2-phenylpentanoyl)oxy]ethyl}azanium bromide |
| SMIL | [Br-].CCC(C)C(C(=O)OCC[N+](C)(CC)CC)C1=CC=CC=C1 |
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiske identifikatorer
| C19H32BrNO2 | |
| MFCD00031617 | |
| valethamate bromide, ediposin, epidosin, epidozin, resitan, murel, valethamate bromide jan, n,n-diethyl-n-methyl-2-3-methyl-2-phenylpentanoyl oxy ethanaminium bromide, diethyl methyl 2-3-methyl-2-phenylvaleryloxy ethyl ammonium bromide, 2-diethylaminoethyl 2-phenyl-3-methylvalerate methyl bromide | |
| diethyl(methyl){2-[(3-methyl-2-phenylpentanoyl)oxy]ethyl}azanium bromide |
Tekniske data
| Valethamate bromide | |
| C19H32BrNO2 | |
| 14,9906 | |
| CEJGGHKJHDHLAZ-UHFFFAOYNA-M | |
| diethyl(methyl){2-[(3-methyl-2-phenylpentanoyl)oxy]ethyl}azanium bromide | |
| 7010 | |
| 99% |
| 90-22-2 | |
| MFCD00031617 | |
| valethamate bromide, ediposin, epidosin, epidozin, resitan, murel, valethamate bromide jan, n,n-diethyl-n-methyl-2-3-methyl-2-phenylpentanoyl oxy ethanaminium bromide, diethyl methyl 2-3-methyl-2-phenylvaleryloxy ethyl ammonium bromide, 2-diethylaminoethyl 2-phenyl-3-methylvalerate methyl bromide | |
| [Br-].CCC(C)C(C(=O)OCC[N+](C)(CC)CC)C1=CC=CC=C1 | |
| 386.37 | |
| 386.37 |
Sikkerhed og håndtering
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'einecsNumber' : 201-977-8
missing translation for 'rtecsNumber' : BP7600000
missing translation for 'tsca' : No
Anbefalet opbevaring : Ambient temperatures
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.