Få mere at vide
Valeric Acid-d9, TRC
Artikelnummer. 30455597
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30455597 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30455597 Leverandør Toronto Research Chemicals Leverandørnr. TRCV091417250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Valeric Acid-d9
CAS 115871-50-6
Molekylær formel C5 D9 H O2
Mængde 250 mg
Synonym 1-Butanecarboxylic Acid-d9, NSC 406833-d9, Propylacetic Acid-d9, n-Pentanoic Acid-d9, n-Valeric Acid-d9, Pentanoic-d9 acid (9CI), Pentanoic acid-d9, Valeric-d9 acid, Pentanoic-D9 acid
InChI formel InChI=1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)/i1D3,2D2,3D2,4D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,3,4,4,5,5,5-nonadeuteriopentanoic acid
Molekylvægt (g/mol) 111.19
Formel vægt 111.1246