Få mere at vide
(-)-Valerenic Acid, TRC
Artikelnummer. 30132853
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132853 5 mg
30003809 10 mg
30006705 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132853 Leverandør Toronto Research Chemicals Leverandørnr. TRCV0914005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3569-10-6
Molekylær formel C15 H22 O2
Molekylvægt (g/mol) 234.33
Synonym (2E)-3-[(4S,7R,7aR)-2,4,5,6,7,7a-hexahydro-3,7-dimethyl-1H-inden-4-yl]-2-methyl-2-propenoic acid, (-)-Valerenic acid, 3-(2,4,5,6,7,7a-hexahydro-3,7-dimethyl-1H-inden-4-yl)-2-methyl-, [4S-[4α(E),7β,7aalpha]]-2-propenoic acid, 2,4,5,6,7,7a-hexahydro-α,3,7-trimethyl-indene-4-acrylic acid, Valerenic acid, 3-(2,4,5,6,7,7a-hexahydro-3,7-dimethyl-1H-inden-4-yl)-2-methyl-, [4S-[4alpha(E),7beta,7aalpha]]-2-propenoic acid, 2,4,5,6,7,7a-hexahydro-alpha,3,7-trimethyl-indene-4-acrylic acid;
IUPAC navn (E)-3-[(4S,7R,7aR)-3,7-dimethyl-2,4,5,6,7,7a-hexahydro-1H-inden-4-yl]-2-methylprop-2-enoic acid
SMIL C[C@@H]1CC[C@@H](\C=C(/C)\C(=O)O)C2=C(C)CC[C@H]12
Tekniske data
Kemisk navn eller materiale Valerenic Acid
Mængde 5 mg
InChI formel InChI=1S/C15H22O2/c1-9-4-6-12(8-11(3)15(16)17)14-10(2)5-7-13(9)14/h8-9,12-13H,4-7H2,1-3H3,(H,16,17)/b11-8+/t9-,12+,13-/m1/s1
Formel vægt 234.162