Få mere at vide
Tocris Bioscience™ Valdecoxib
GREENER_CHOICE
Artikelnummer. 18023932
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18023932 10 mg
18055542 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18023932 Leverandør Tocris Bioscience™ Leverandørnr. 4206/10
Selective and potent COX-2 inhibitor
Selective and potent COX-2 inhibitor (in vitro IC50 values are 0.005 and 140 μM for human recombinant COX-2 and COX-1 respectively). Displays potent anti-inflammatory activity in vivo.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 181695-72-7
Molekylær formel C16H14N2O3S
Tekniske data
Kemisk navn eller materiale Valdecoxib
Mængde 10 mg
Formel vægt 314.36
Renhed >99%
Molekylvægt (g/mol) 314.359