Få mere at vide
Valdecoxib-d3 (methyl-d3), CDN
Artikelnummer. 30311815
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311815 0.01 g
30322376 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311815 Leverandør CDN Leverandørnr. CDND68410.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Valdecoxib methyl-d3
CAS 1219794-90-7
Molekylær formel C16 2H3 H11 N2 O3 S
Mængde 0.01 g
InChI formel InChI=1 S/C16H14N2O3S/c1-11-15(12-7-9-14(10-8-12)22(17,19)20)16(18-21-11)13-5-3-2-4-6-13/h2-10 H,1H3,(H2,17,19,20)/i1D3
SMIL [2 H]C([2 H])([2 H])c1onc(c2ccccc2)c1c3ccc(cc3)S(=O)(=O)N
IUPAC navn 4-[3-phenyl-5-(trideuteriomethyl)-1,2-oxazol-4-yl]benzenesulfonamide
Molekylvægt (g/mol) 317.38
Formel vægt 317.0913 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity