Learn More
Thermo Scientific™ Valco™ Injector Model C6W
Use the Thermo Scientific™ Valco™ Injector Model C6W as an injector or switching valve.
Brand: Thermo Scientific™ SL5CW
Description
Use the Thermo Scientific™ Valco™ Injector Model C6W as an injector or switching valve. Available with manual or a microelectric actuator. Has 1/16 in. fittings and six 0.010 inch ports.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Microelectric or Manual | |
Sample injector loop, stainless steel | |
6 Ports |
5 ÎĽL | |
Injector/Switching Valve | |
Stainless Steel |