Få mere at vide
Valacyclovir-d4, Hydrochloride, TRC
Artikelnummer. 30454471
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30454471 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454471 Leverandør Toronto Research Chemicals Leverandørnr. TRCV08500210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Valacyclovir-d4','Hydrochloride
CAS 1331910-75-8
Molekylær formel C13H17D4ClN6O4
Mængde 10 mg
Synonym L-Valine 2-[(2-Amino-1,6-dihydro-6-oxo-9H-purin-9yl)methoxy]ethyl Ester-d4','Hydrochlroride Salt','ValACV
InChI formel InChI=1S/C13H20N6O4.ClH/c1-7(2)8(14)12(21)23-4-3-22-6-19-5-16-9-10(19)17-13(15)18-11(9)20;/h5,7-8H,3-4,6,14H2,1-2H3,(H3,15,17,18,20);1H/t8-;/m0./s1/i3D2,4D2;
SMIL O=C1C2=C(N(COC([2H])([2H])C([2H])([2H])OC([C@@H](N)C(C)C)=O)C=N2)N=C(N)N1.Cl
Molekylvægt (g/mol) 364.82
Formel vægt 364.1564