Få mere at vide
Valaciclovir Hydrochloride Monohydrate, LoGiCal
Artikelnummer. 30458522
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30458522 10 mg
1 options
Artikelnummer. 30458522 Leverandør LoGiCal Leverandørnr. LGCFOR3012.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Valaciclovir Hydrochloride Monohydrate
CAS 521915-75-3
Analyt- eller komponentnavne Valaciclovir Hydrochloride Monohydrate
Forsendelsestilstand Room Temperature
Molekylær formel C13 H20 N6 O4 . Cl H . H2 O
Certificeringer/compliance ISO 9001
Synonym Valaciclovir hydrochloride monohydrate, L-Valine, 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]ethyl ester, monohydrochloride, monohydrate
InChI formel InChI=1S/C13H20N6O4.ClH.H2O/c1-7(2)8(14)12(21)23-4-3-22-6-19-5-16-9-10(19)17-13(15)18-11(9)20;;/h5,7-8H,3-4,6,14H2,1-2H3,(H3,15,17,18,20);1H;1H2/t8-;;/m0../s1
Løsningstype Neat
SMIL O.Cl.CC(C)[C@H](N)C(=O)OCCOCn1cnc2C(=O)NC(=Nc12)N
IUPAC navn 2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]ethyl (2S)-2-amino-3-methylbutanoate;hydrate;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 378.81
Mængde 10 mg
Formel vægt 378.1418 g/mol