Learn More
Greiner Bio-One Vacuette Tube™, Lavender Cap-White Ring, K2E K2EDTA, Sterile
Product Code. 15402530
Change view
Click to view available options
Capacity (Metric):
2 mL
Closure Color:
Lavender with White Ring
Unit Size:
Pack of 50
Product Code. Capacity (Metric) Closure Color Unit Size
15402530 2 mL Lavender with White Ring Pack of 50
1 options
You may only order up to the available stock. For more details call customer services on +45 70 27 99 20.
Product Code. 15402530 Supplier Greiner Bio-One Supplier No. 454047
  • ONLINE EXCLUSIVE PRICE
    128.10 DKK
    75.62 DKK / Pakke med 50
    Spare 52.48 DKK
    40% Slukket
 Restricted Item
Forventet forsendelse: 23-12-2026
You may only order up to the available stock. For more details call customer services on +45 70 27 99 20.
Inner wall is coated with anticoagulant that binds calcium ions and interrupts clotting.
  • Inner wall is coated with spray-dried K2E K2EDTA as an anticoagulant to bind calcium ions and interrupt the clotting cascade.
  • Inner wall is coated with spray-dried K2E K2EDTA as an anticoagulant to bind calcium ions and interrupt the clotting cascade.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vacuette Tube
Additive K2 E K2 EDTA
Sterility Sterile
Capacity (Metric) 2 mL
Closure Color Lavender with White Ring
Length (Metric) 75 mm
Diameter (Metric) Outer 13 mm