Få mere at vide
Vaccenic Acid Ethyl Ester, TRC
Artikelnummer. 30455482
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30455482 25 mg
30449784 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30455482 Leverandør Toronto Research Chemicals Leverandørnr. TRCV07000525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Vaccenic Acid Ethyl Ester
CAS 89296-48-0
Molekylær formel C20H38O2
Mængde 25 mg
Synonym 11-Octadecenoic Acid Ethyl Ester, (E)-11-Octadecenoic Acid Ethyl Ester, Ethyl Vaccenate
InChI formel InChI=1S/C20H38O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h9-10H,3-8,11-19H2,1-2H3/b10-9+
SMIL O=C(OCC)CCCCCCCCC/C=C/CCCCCC
IUPAC navn ethyl (E)-octadec-11-enoate
Molekylvægt (g/mol) 310.51
Formel vægt 310.29