missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ V5 Tag Monoclonal Antibody (A10-D0-A1)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA544633
This item is not returnable.
View return policy
Description
Application notes: Indirect ELISA.
Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. The V5 epitope tag is derived from a small epitope (Pk) present on the P and V proteins of the paramyxovirus of simian virus (SV5). The V5 tag is usually used with all 14 amino acids (GKPIPNPLLGLDST), although it has also been used with a shorter 9 amino acid sequence (IPNPLLGLD).
Specifications
V5 Tag | |
Monoclonal | |
2 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Tag | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
A10-D0-A1 | |
Unconjugated | |
simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag | |
Peptide GKPIPNPLLGLDST (V5 epitope) conjugated with KLH. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ V5 Tag Monoclonal Antibody (A10-D0-A1) > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction