Få mere at vide
V23 Wear Metals Standard: 50 μg/g Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG™
Artikelnummer. 16419679
Skift visning
Mængde:
100 g
200 g
400 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16419679 100 g
17918269 200 g
17983916 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16419679 Leverandør VHG Labs Leverandørnr. VHGV2350100G
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
V23 Wear Metals Standard: 50 μg/g Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Aluminium/Antimony/Barium/Boron/Cadmium/Calcium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc
CAS 7429-90-5, 7440-36-0, 7440-39-3, 7440-42-8, 7440-43-9, 7440-70-2
Metoder ICP/XRF/RDE
Analyt- eller komponentnavne Aluminium/Antimony/Barium/Boron/Cadmium/Calcium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc
Forsendelsestilstand Room Temperature
Certificeringer/compliance ISO 17034
Opløsningsmiddel eller Matrix Hydrocarbon Oil
Mængde 100 g
Koncentration eller sammensætning (efter analyt eller komponenter) Aluminium 50 μg/g/Antimony 50 μg/g/Barium 50 μg/g/Boron 50 μg/g/Cadmium 50 μg/g/Calcium 50 μg/g/Chromium 50 μg/g/Copper 50 μg/g/Iron 50 μg/g/Lead 50 μg/g/Magnesium 50 μg/g/Manganese 50 μg/g/Molybdenum 50 μg/g/Nickel 50 μg/g/Phosphorus 50 μg/g/Potassium 50 μg/g/Silicon 50 μg/g/Silver 50 μg/g/Sodium 50 μg/g/Tin 50 μg/g/Titanium 50 μg/g/Vanadium 50 μg/g/Zinc 50 μg/g