Learn More
Uvex™ Polycarbonate Lens Safety Glasses
Catalog Number p-8000266
This item is not returnable. View return policy
This item is not returnable. View return policy
Adjustable safety spectacles
Innovative styling and design, with adjustable temple length and lens inclination.
  • Wraparound style spectacle with excellent field of vision
  • Straight side arms grip the side of the head not behind the ears. Provides a secure comfortable, slip free wear
  • Combines hard/soft components - increased comfort around sensitive nose, brow and ear areas
  • Fully adjustable side arms enables wearer to achieve individual fit for better protection and comfort
  • 100% UV protection and metal free
  • Clear lens – suitable for most light environments
  • Clear/2-1.2
  • Blue/Orange
Lens Tint Clear
Product Type Glasses; Sports Style
Frame Color Blue, Orange
No. per Pack 1 Ea.
Temple Style Adjustable
Lens Material Polycarbonate