Få mere at vide
Honeywell™ Uvex™ Genesis™ Protective Safety Glasses
Artikelnummer. 12327289
Skift visning
Ramme farve:
Vapor Blue
Linsebelægning:
Anti-fog
Linsefarve:
Clear
Pakningsstørrelse:
Hver
Artikelnummer. Ramme farve Linsebelægning Linsefarve unitSize
12327289 Vapor Blue Anti-fog Clear Hver
1 options
Artikelnummer. 12327289 Leverandør Honeywell™ Leverandørnr. 19040169
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Three-position ratchet lens inclination system and adjustable temples enable wearer to customize fit
  • Impact-resistant polycarbonate lenses block >99.9% of harmful UVA and UVB radiation
  • Ventilation channel between frame and lens prevents moisture buildup
  • Nose bridge is covered with small, ultrasoft “fingers” that hug the nose for improved fit and comfort
  • Soft elastomer browguard cushions all points of contact with face
uvextreme™ Anti-Fog Coating (XTR)
  • Permanently bonded to lens
  • Absorbs 99.9% of UV radiation
  • Anti-scratch and -static
  • Will not wear after repeated cleanings
Hydroshield™ Anti-Fog Coating
  • Provides 99.9% ultra-violet protection
  • Provides 2X greater scratch-resistance
XC Series
  • Extended wrap-around lens for extra cheek and extra side protection
    • Soft flexible nose fingers conform to fit a variety of nasal profiles
    • Adjustable temples and lens inclination
    ultra-dura™ Hard Coat (UD)
    • Permanently bonded to lens
    • Scratch- and chemical-resistant
Overholdelse
Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards. Meet stringent military V0 Ballistic Test for Impact requirements.
garanti
Lifetime frame guarantee.
TRUSTED_SUSTAINABILITY
Tekniske data
Certificeringer/compliance ANSI Z87.1-2015, CSA Z94.3-2007, Military V0 Ballistic Test
Linsebelægning Anti-fog
Oversigt Lifetime frame guarantee
Linsefarve Clear
Ramme farve Vapor Blue