Få mere at vide
Uvex™ Astrospec NC
Artikelnummer. 10543211 Shop alle Uvex produkter
:
Frame Color: Blue
Artikelnummer. 10543211
Brand: Uvex™ 9168065
Pleasantly lightweight single lens with integrated side protection
  • Pleasantly lightweight single lens with integrated side protection
  • Individual strap inclination and length adjustment uvex duo-flex strap, easy disc change
  • Frame color blue, W 166 F CE
  • Disc colorless
  • UV 400, 2-1,2 W 1 F DIN CE
  • Coating uvex supravision sapphire
  • Spare disc: 9168.055
  • TRUSTED_SUSTAINABILITY
    Tekniske data
    Certificeringer/compliance EN 170
    Linsetype Chemical-resistant
    Linsebelægning Scratch-resistant
    Oversigt Strap glasses plastic
    Linsefarve Clear
    Ramme farve Blue