Learn More
Fisherbrand™ Gel Casting Tray
Product Code. 11573382 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Wide Format Midi-Plus Horizontal Gel Unit
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
11573382 Wide Format Midi-Plus Horizontal Gel Unit Each
1 options
Product Code. 11573382
  • ONLINE EXCLUSIVE PRICE
    888.00 DKK
    850.77 DKK / Pakke med 1
    Spare 37.23 DKK
    4% Slukket
Forventet forsendelse: 03-02-2027
Add to basket
Add to basket
Accessory for Wide Format Midi-Plus Horizontal Gel Units.
  • Measures 100 x 230mm
Specifications
For Use With (Equipment) Wide Format Midi-Plus Horizontal Gel Unit
Type Gel Casting Tray
Dimensions (L x W) 100 x 230 mm