Få mere at vide
UTP3 Antibody, CoraFluor™ 1, Novus Biologicals™
Artikelnummer. 30141586 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mL
Pakningsstørrelse:
0.1ml
Artikelnummer. Mængde unitSize
30141586 0.1 mL 0.1ml
1 options
Artikelnummer. 30141586 Leverandør Novus Biologicals Leverandørnr. NBP119108CL1
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Rabbit Polyclonal Antibody
UTP3 Polyclonal antibody specifically detects UTP3 in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen UTP3
Applikationer Immunoprecipitation, Western Blot
Klassifikation Polyclonal
Konjugeret CoraFluor 1
Formulering PBS
Gene Alias Charged amino acid-rich leucine zipper 1, CRL1charged amino acid rich leucine zipper 1 homolog, CRLZ1UTP3 homolog, disrupter of silencing 10, Disrupter of silencing SAS10, FLJ23256, SAS10DKFZp761F222, something about silencing protein 10, UTP3, small subunit (SSU) processome component, homolog (S. cerevisiae)
Værtsarter Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 429 and 479 of human something about silencing protein 10 using the numbering given in entry NP_065101.1 (GeneID 57050).
Oprensningsmetode Immunogen affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Chromatin Modifiers
Primær eller sekundær Primary
Gen-id (Entrez) 57050
Målarter Human
Indhold og opbevaring Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG