missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Uteroglobin/SCGB1A1 Antibody (027), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90516V
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Uteroglobin/SCGB1A1 Monoclonal specifically detects Uteroglobin/SCGB1A1 in Mouse samples. It is validated for ELISA.
Tekniske data
| Uteroglobin/SCGB1A1 | |
| Monoclonal | |
| DyLight 405 | |
| blastokinin, CC10Urine protein 1, CC16, CCSPCCPBP, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGBUP-1, UP1, Urinary protein 1, uteroglobin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Uteroglobin/SCGB1A1 (Q06318; Met1-Phe96). | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 7356 | |
| Mouse | |
| Purified |
| ELISA | |
| 27 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human SCGB1A1 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion