Få mere at vide
USP16 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-81638AF532
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
USP16 Polyclonal antibody specifically detects USP16 in Human samples. It is validated for Western Blot
Tekniske data
| USP16 | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| 0.1 mL | |
| 10600 | |
| Purified |
| Polyclonal | |
| Western Blot | |
| Deubiquitinating enzyme 16, EC 3.1.2.15, EC 3.4.19.12, human ubiquitin processing protease, EC 3.1.2.1510ubiquitin specific protease 16, ubiquitin carboxyl-terminal hydrolase 16, ubiquitin specific peptidase 16, ubiquitin thioesterase 16, Ubiquitin thiolesterase 16, Ubiquitin-processing protease UBP-M, ubiquitin-specific processing protease 16, Ubiquitin-specific-processing protease 16, UBP-M | |
| Cell Cycle and Replication | |
| Store at 4C in the dark. | |
| IgG |