Få mere at vide
USL311, MedChemExpress
Artikelnummer. 16494798
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16494798 10 mg
16457547 5 mg
16451067 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16494798 Leverandør Medchem Express Leverandørnr. HY114244
MedChemExpress USL311 is a selective CXCR4 antagonist, with anti-tumor activity. USL311 prevents the binding of stromal-cell derived factor-1 (SDF-1 or CXCL12) to CXCR4.
USL311 is a selective CXCR4 antagonist, with anti-tumor activity. USL311 prevents the binding of stromal-cell derived factor-1 (SDF-1 or CXCL12) to CXCR4.
Tekniske data
Kemisk navn eller materiale USL311
CAS 1373268-67-7
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C24H34N6O
Mængde 10 mg
Opløselighedsinformation DMSO : 18.57 mg/mL (43.95 mM; ultrasonic and warming and heat to 80°C)
SMIL O=C(C1=NC(N2CCN(C3CCN(C(C)C)CC3)CCC2)=CC=C1)NC4=CC=NC=C4
Molekylvægt (g/mol) 422.57
Formel vægt 422.57
Procent renhed 99.78%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death