missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ USB-Ethernet Adapter for UN160
USB-Ethernet Adapter accessory for Memmert Ovens.
Brand: Memmert™ UN160E06192
This item is not returnable.
View return policy
Description
- Accessory
- For use with Memmert Ovens
Specifications
UN160 Oven | |
USB Ethernet Adapter |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ USB-Ethernet Adapter for UN160 > For Use With: UN160
Spot an opportunity for improvement?Share a Content Correction