Få mere at vide
Ursodeoxycholic Acid-d4 (Major), TRC
Artikelnummer. 30451537
Skift visning
Mængde:
1 mg
2.5 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30451537 1 mg -
30450412 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451537 Leverandør Toronto Research Chemicals Leverandørnr. TRCU8500031MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ursodeoxycholic-2,2,4,4-d4 acid
CAS 347841-46-7
Molekylær formel C24 2H4 H36 O4
Mængde 1 mg
Synonym Cholan-24-oic-2,2,4,4-d4 acid','3,7-dihydroxy-','(3α,5β,7β)-, (3α,5β,7β)-3,7-Dihydroxycholan-24-oic-2,2,4,4-d4 acid, Ursodeoxycholic acid-d4, Ursodesoxycholic-2,2,4,4-d4 acid, Ursodeoxycholic-2,2,4,4-D4 acid
InChI formel InChI=1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20+,22+,23+,24-/m1/s1/i8D2,12D2
SMIL [2H]C1([2H])C[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3[C@@H](O)C[C@@H]2C([2H])([2H])[C@@H]1O)[C@H](C)CCC(=O)O
IUPAC navn (4R)-4-[(3R,5R,7S,8R,9S,10S,13R,14S,17R)-2,2,4,4-tetradeuterio-3,7-dihydroxy-10,13-dimethyl-3,5,6,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molekylvægt (g/mol) 396.6
Formel vægt 396.3178