Få mere at vide
Ursodeoxycholic-2,2,4,4-d4 Acid, CDN
Artikelnummer. 30325027
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30325027 0.05 g -
30323817 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325027 Leverandør CDN Leverandørnr. CDND38190.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ursodeoxycholic-2,2,4,4-d4 acid
CAS 347841-46-7
Molekylær formel C24 2H4 H36 O4
Mængde 0.05 g
Synonym Cholan-24-oic-2,2,4,4-d4 acid, 3,7-dihydroxy-, (3α,5β,7β)-, (3α,5β,7β)-3,7-Dihydroxycholan-24-oic-2,2,4,4-d4 acid, Ursodeoxycholic acid-d4, Ursodesoxycholic-2,2,4,4-d4 acid, Ursodeoxycholic-2,2,4,4-D4 acid
InChI formel InChI=1 S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26 H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20+,22+,23+,24-/m1/s1/i8D2,12D2
SMIL [2 H]C1([2 H])C[C@]2(C)[C@H]3 CC[C@]4(C)[C@H](CC[C@H]4 [C@@H]3 [C@@H](O)C[C@@H]2 C([2 H])([2 H])[C@@H]1 O)[C@H](C)CCC(=O)O
IUPAC navn (4 R)-4-[(3 R,5 R,7 S,8 R,9 S,10 S,13 R,14 S,17 R)-2,2,4,4-tetradeuterio-3,7-dihydroxy-10,13-dimethyl-3,5,6,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molekylvægt (g/mol) 396.6
Formel vægt 396.3178 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity