Få mere at vide
URMC-099, MedChemExpress
Artikelnummer. 16495258
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16495258 10 mg 10mg
16457977 5 mg 5mg
16445926 100 mg -
16401477 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16495258 Leverandør Medchem Express Leverandørnr. HY12599
MedChemExpress URMC-099 is an orally bioavailable and potent mixed lineage kinase type 3 (MLK3) (IC50=14 nM) inhibitor with with excellent blood-brain barrier penetration properties.
URMC-099 is an orally bioavailable and potent mixed lineage kinase type 3 (MLK3) (IC50=14 nM) inhibitor with with excellent blood-brain barrier penetration properties.
Tekniske data
Kemisk navn eller materiale URMC-099
CAS 1229582-33-5
Farve Light Nude
Renhedsgrad noter Research
Molekylær formel C27H27N5
Mængde 10 mg
Opløselighedsinformation DMSO : ≥ 33 mg/mL (78.28 mM)
SMIL CN(CC1)CCN1CC(C=C2)=CC=C2C3=CN=C4C(C(C5=CC=C(NC=C6)C6=C5)=CN4)=C3
Molekylvægt (g/mol) 421.54
Formel vægt 421.54
Procent renhed 98.02%
Grad Research
Fysisk form Powder
Til brug med (applikation) Cancer-Kinase/protease