missing translation for 'onlineSavingsMsg'
Learn More

URM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1081.84 DKK - 3058.99 DKK

Specifications

Antigen URM1
Concentration 0.46 mg/mL
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16814404
View Documents
Proteintech
15285-1-AP-20UL
20 ÎĽL
1081.84 DKK
20µL
Please to purchase this item. Need a web account? Register with us today!
16804404
View Documents
Proteintech
15285-1-AP-150UL
150 ÎĽL
3058.99 DKK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Following the discovery of protein modification by the small, highly conserved ubiquitin polypeptide, a number of distinct ubiquitin-like proteins (Ubls) have been found to function as protein modifiers as well. These Ubls, which include SUMO, ISG15, Nedd8, and Atg8, function as critical regulators of many cellular processes, including transcription, DNA repair, signal transduction, autophagy, and cell-cycle control. A growing body of data also implicates the dysregulation of Ubl-substrate modification and mutations in the Ubl-conjugation machinery in the etiology and progression of a number of human diseases. URM1 is a ubiquitin-like modifier protein.
Specifications

Specifications

URM1
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Rat, Mouse
Q9BTM9, Q9D2P4
311840, 68205, 81605
URM1 Fusion Protein Ag7448
Primary
-20°C
URM1
0.46 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
C9orf74, URM1
Urm1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
URM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.