Learn More
Uridine, ∽99%, MP Biomedicals™ Product Code.: 11427441
Uridine, ∽99%, MP Biomedicals™
BIOPHARMA
Product Code. 11427441 Shop All MP Biomedical Products
100 g, 100g
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
25g
5g
Product Code. 11427441
Brand: MP Biomedicals 0210321680
Chemical Identifiers
CAS 58-96-8
Molecular Formula C9H12N2O6
Molecular Weight (g/mol) 244.203
InChI Key DRTQHJPVMGBUCF-DHRBYNEYSA-N
Synonym uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd
PubChem CID 89495556
IUPAC Name 1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O
Specifications
Color White
Assay Percent Range ∽99%
Quantity 100 g
Formula Weight 244.2
Physical Form Powder
Chemical Name or Material Uridine
Product Content Correction
Product Title
Uridine, ∽99%, MP Biomedicals™ >