Learn More
Uridine, ∽99%, MP Biomedicals™ Product Code.: 11417441
Uridine, ∽99%, MP Biomedicals™
BIOPHARMA
Product Code. 11417441 Shop All MP Biomedical Products
25 g, 25g
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
25g
5g
Product Code. 11417441
Brand: MP Biomedicals 0210321625
Chemical Identifiers
CAS 58-96-8
Molecular Formula C9H12N2O6
Molecular Weight (g/mol) 244.203
InChI Key DRTQHJPVMGBUCF-DHRBYNEYSA-N
Synonym uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urdShow MoreShow Less
PubChem CID 89495556
IUPAC Name 1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O
Specifications
Color White
Assay Percent Range ∽99%
Quantity 25 g
Formula Weight 244.2
Physical Form Powder
Chemical Name or Material Uridine
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Product Code
Lot NumberU1123005265 Certificate TypeCertificate of Analysis Date24/03/2023 Product Code0210321625
Lot NumberU1123005264 Certificate TypeCertificate of Analysis Date24/03/2023 Product Code0210321625
Lot NumberU1122284481 Certificate TypeCertificate of Analysis Date24/03/2023 Product Code0210321625
Lot NumberS9303 Certificate TypeCertificate of Analysis Date24/10/2022 Product Code0210321625
Product Content Correction
Product Title
Uridine, ∽99%, MP Biomedicals™ >