missing translation for 'onlineSavingsMsg'
Få mere at vide

Uridin 5'-Diphosphat Natriumsalt, TRC

Artikelnummer. 30455426
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30455426 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30455426 Leverandør Toronto Research Chemicals Leverandørnr. TRCU8299201G
 Begrænset vare
Forventet forsendelse: 03-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Uridine 5'-Diphosphate Sodium Salt
CAS 21931-53-3
Molekylær formel C9 H12 N2 O12 P2 . 2 Na
Mængde 1 g
Synonym Uridine 5'-Diphosphate Sodium Salt
InChI formel InChI=1S/C9H14N2O12P2.2Na/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(22-8)3-21-25(19,20)23-24(16,17)18;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,10,12,15)(H2,16,17,18);;/q;2*+1/p-2/t4-,6-,7-,8-;;/m1../s1
SMIL [Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)([O-])OP(=O)(O)[O-])N2C=CC(=O)NC2=O
IUPAC navn disodium; [[(2R,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogenfosfat
Molekylvægt (g/mol) 448.12
Formel vægt 447.9661
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.