Få mere at vide
Uridine-5,6-d2, TRC
Artikelnummer. 30452631
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30452631 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452631 Leverandør Toronto Research Chemicals Leverandørnr. TRCU8299125MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Uridine-5,6-d2
CAS 40632-21-1
Molekylær formel C9 2H2 H10 N2 O6
Mængde 5 mg
Synonym Uridine-5,6-d2 (9CI), 5,6-Dideuteriouridine, Uridine-5,6-D2, [5,6-2H2]uridine, 1-β-D-Ribofuranosyl-2,4(1H,3H)-pyrimidinedione-5,6-d2, 1-β-D-Ribofuranosyluracil-5,6-d2
InChI formel InChI=1S/C9H12N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h1-2,4,6-8,12,14-15H,3H2,(H,10,13,16)/t4-,6-,7-,8-/m1/s1/i1D,2D
SMIL [2H]C1=C([2H])C(=O)NC(=O)N1[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O
IUPAC navn 5,6-dideuterio-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
Molekylvægt (g/mol) 246.21
Formel vægt 246.0821