Få mere at vide
Uridine-2-13C-1,3-15N2, TRC
Artikelnummer. 30010567
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010567 10 mg
30009816 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30010567 Leverandør Toronto Research Chemicals Leverandørnr. TRCU82990710MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 369656-75-7
Molekylær formel 13C C8 H12 15N2 O6
Molekylvægt (g/mol) 247.18
Synonym Uridine-2-13C-1,3-15N2 (9CI), [2-13C-1,3-15N2]Uridine, 1-β-D-Ribofuranosyl-2,4(1H,3H)-pyrimidinedione-2-13C-1,3-15N2, 1-β-D-Ribofuranosyluracil-2-13C-1,3-15N2
IUPAC navn 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl](2^{13}C,1,3-^{15}N_{2})pyrimidine-2,4-dione
SMIL OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)[15N]2C=CC(=O)[15NH][13C]2=O
Tekniske data
Kemisk navn eller materiale Uridine-2-13C-1,3-15N2
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C9H12N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h1-2,4,6-8,12,14-15H,3H2,(H,10,13,16)/t4-,6-,7-,8-/m1/s1/i9+1,10+1,11+1
Formel vægt 247.067
Procent renhed >95%